CD57 Mouse Monoclonal Antibody (clone NK-1), Concentrate

Catalog number: 180167Z

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


This monoclonal antibody reacts with CD57, a 110 kDa human lymphocyte cell surface antigen. Labels Natural Killer cells (large granular lymphocytes) in germinal centers. The antibody is useful in the detection of large granular lymphocytosis. The antigen is expressed in 50% of small cell lung carcinomas and 85% of carcinoid tumors. Positive tissue: Tonsil/lymph node/spleen.

Clone/PAD: NK-1

Immunogen: Lymph node cell suspensions

Specificity: CD57

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (Citrate Buffer pH 6.0) OR Enzyme Digestion: Required (Digest-All™ 2/Trypsin)

Expected Staining Pattern: Cytoplasm and membrane
For In Vitro Diagnostic Use.


Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
SubClass (Isotype): IgM
Clonality: Monoclonal
Target Gene: CD57
Gene ID: 27087
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).


Manuals & protocols