Cytokeratin 8 Mouse Monclonal Antibody (clone C51), Concentrate

Catalog number: 180185Z

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


This monoclonal antibody reacts with human cytokeratin 8. Positive tissue: Colon/gastric mucosa.

Clone/PAD: C51

Immunogen: Cytokeratin preparation form HeLa.

Specificity: Reacts with human cytokeratin 8 (52.5 kDa). Antibody has no cross-reactivity to other filament types, including vimentin, desmin, glial fibrillary acidic protein and neurofilament protein.

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (Citrate Buffer pH 6.0) OR Enzyme Digestion: (Digest-All™ 3/Pepsin)
For In Vitro Diagnostic Use.


Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
SubClass (Isotype): IgG1, kappa
Clonality: Monoclonal
Target Gene: Cytokeratin 8
Gene ID: 3856
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).


Manuals & protocols