Ribonuclease H (RNase H) is an endoribonuclease that specifically degrades the RNA strand of an RNA-DNA hybrid to produce 5´Read more
Have Questions?
Change view
Catalog Number
Quantity
18021014
30 U
18021071
120 U
2 Options
Catalog number 18021014
Price (USD)
232.65
Online exclusive
256.00
Save 23.35
Each
Quantity:
30 U
Request bulk or custom format
Price (USD)
232.65
Online exclusive
256.00
Save 23.35
Each
Ribonuclease H (RNase H) is an endoribonuclease that specifically degrades the RNA strand of an RNA-DNA hybrid to produce 5´ phosphate-terminated oligoribonucleotides and single-stranded DNA. Applications Removal of mRNA during second-strand cDNA synthesis. Removal of poly(A) sequences from mRNA in the presence of oligo(dT). Oligodeoxyribonucleotide-directed cleavage of RNA. Source Purified from E. coli expressing the E. coli RNase H gene on a plasmid. Performance and quality testing Ribonuclease, nonspecific endodeoxyribonuclease, 3´ and 5´ exodeoxyribonuclease. Unit definition One unit hydrolyzes 1 nmol of RNA in 3H-labeled poly(A)poly(dT) to acid-soluble material in 20 min at 37°C. Unit reaction conditions 20 mM Tris-HCl (pH 7.5), 0.1 M KCl, 10 mM MgCl2 , 0.1 mM DTT, 5% (w/v) sucrose, 0.5 nmol 3H-labeled poly(A)poly(dT), and enzyme in 50 μL for 20 min at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeRibonuclease H
Quantity30 U
Shipping ConditionApproved for shipment on Wet or Dry Ice
Concentration2 U/μL
EnzymeRNase
Unit SizeEach
Contents & Storage
1 vial of Ribonuclease H containing 30 units supplied at a concentration of 2U/μl
Store at -20°C in a non-frost-free freezer.
Frequently asked questions (FAQs)
Do you offer RNAse H from the SuperScript III First Strand Synthesis System as a stand-alone item?
Yes, we do offer E. coli RNAse H as a stand-alone item: Cat. Nos. 18021014 and 18021071.
Which components of the SuperScript III First Strand Synthesis System for RT-PCR are available for purchase separately?
The following components are available as stand-alone items:
- Superscript III Reverse Transcriptase (Cat. Nos. 18080093, 18080044, 18080085)
- Oligo (dT)20 Primer (Cat. No. 18418020)
- Random hexamers (Cat. No. 48190011)
- 10 mM dNTP Mix (Cat. Nos. 18427013, 18427088)
- RNAseOUT Recombinant Ribonuclease Inhibitor (Cat. No. 10777019)
- E. coli RNAse H (Cat. Nos. 18021014, 18021071)
Citations & References (6)
Citations & References
Abstract
Functions of Transforming Growth Factor-beta Family Type I Receptors and Smad Proteins in the Hypertrophic Maturation and Osteoblastic Differentiation of Chondrocytes.
'We investigated the effects of bone morphogenetic protein (BMP)-2, a member of the transforming growth factor-beta superfamily, on the regulation of the chondrocyte phenotype, and we identified signaling molecules involved in this regulation. BMP-2 triggers three concomitant responses in mouse primary chondrocytes and chondrocytic MC615 cells. First, BMP-2 stimulates expression ... More
De novo ceramide regulates the alternative splicing of caspase 9 and Bcl-x in A549 lung adenocarcinoma cells. Dependence on protein phosphatase-1.
Authors: Chalfant Charles E; Rathman Kristin; Pinkerman Ryan L; Wood Rachel E; Obeid Lina M; Ogretmen Besim; Hannun Yusuf A;
Journal:J Biol Chem
PubMed ID:11801602
'In a recent study, we showed that ceramide induces the dephosphorylation of SR proteins, a family of protein factors that regulate alternative splicing. In this study, the regulation of the alternative processing of pre-mRNA of both caspase 9 and Bcl-x(L) was examined in response to ceramide. Treatment of A549 lung ... More
The novel WD-repeat protein MORG1 acts as a molecular scaffold for HIF prolyl-hydroxylase 3 (PHD3).
Authors:Hopfer U, Hopfer H, Jablonski K, Stahl RA, Wolf G,
Journal:J Biol Chem
PubMed ID:16407229
'Hypoxia-inducible factor-1 (HIF-1), a transcriptional complex composed of an oxygen-sensitive alpha- and a beta-subunit, plays a pivotal role in cellular adaptation to low oxygen availability. Under normoxia, the alpha-subunit of HIF-1 is hydroxylated by a family of prolyl hydroxylases (PHDs) and consequently targeted for proteasomal degradation. Three different prolyl hydroxylases ... More
High-sensitivity detection of DNA hybridization on microarrays using resonance light scattering.
Authors: Bao Paul; Frutos Anthony G; Greef Charles; Lahiri Joydeep; Muller Uwe; Peterson Todd C; Warden Laurence; Xie Xinying;
Journal:Anal Chem
PubMed ID:11985309
The application of resonance light scattering (RLS) particles for high-sensitivity detection of DNA hybridization on cDNA microarrays is demonstrated. Arrays composed of approximately 2000 human genes ( ... More
mRNA decay is rapidly induced after spore germination of Saccharomyces cerevisiae.
Spores from the yeast Saccharomyces cerevisiae can germinate and resume their vegetative growth when placed in favorable conditions. Biochemical studies on germination have been limited by the difficulty of obtaining a pure population of spores germinating synchronously. Here, we report that spores can be purified and sorted according to their ... More