EGP2 (Epithelial Glycoprotein 2) Mouse Monoclonal Antibody (clone MOC-31), Concentrate

Catalog number: 180270

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with EGP-2. In normal tissues, clone MOC-31 detects a membrane epithelial antigen and is immunoreactive with pancreas, prostate, brain, kidney, thyroid gland and colon. In tumors, clone MOC-31 reacts with both SCLC and non-SCLC. Clone MOC-31 is usually non-reactive in normal and malignant mesothelia and therefore is especially useful for the distinction between lung carcinomas and mesotheliomas. MOC-31 stains adenocarcinoma cells but not reactive mesothelial cells in pleural effusions. Positive tissue: Small cell lung cancer.

Clone/PAD: MOC-31

Immunogen: Cells derived from small cell lung carcinoma

Specificity: Epithelial glycoprotein 2 (EGP2)

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Not Required, Enzyme Digestion: Required [Digest-All 3/Pepsin)

Expected Staining Pattern: Membrane

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
SubClass (Isotype): IgG1
Clonality: Monoclonal
Target Gene: EGP2 (Epithelial Glycoprotein 2)
Gene ID: 4072
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products