CO<sub>2</sub> Independent Medium
Gibco™

CO2 Independent Medium

CO2 Independent Medium is a non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherentRead more
Catalog NumberQuantity
18045088500 mL
Catalog number 18045088
Price (JPY)
12,700
Each
Quantity:
500 mL

CO2 Independent Medium is a non-HEPES proprietary medium used for supporting cell growth for a variety of suspension and adherent mammalian cells such as epithelial, fibroblast, and lymphoid cell lines without a CO2 incubator. CO2 Independent Medium is ideal for transporting cells or tissue and for the handling of mouse embryos under atmospheric conditions.

This CO2 Independent Medium is manufactured as follows:

  • With
    • Phenol Red
    • Sodium pyruvate
  • Without
    • L-glutamine
    • HEPES

The complete formulation is confidential. For more information, please contact Technical Support.

Gibco™ CO2 Independent Medium contains a unique buffering system composed of mono and dibasic sodium phosphate and β-glycerophosphate. A small amount of sodium bicarbonate has been included in the formulation to meet essential bicarbonate dependent functions. No synthetic buffers are utilized, thus eliminating any cytotoxic effects associated with such buffering systems. Additionally, Gibco™ CO2 Independent Medium has been formulated with components that enhance cellular production and utilization of CO2 such that an exogenous source of CO2 is not required for the maintenance of CO2 dependent cellular functions.

Product Use
For research use only: Not intended for human or animal diagnostic or therapeutic uses.

Dual-Site cGMP Manufacturing
Gibco™ CO2 Independent Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, Life Technologies offers an identical Gibco™ CO2 Independent Medium product made in our Scotland facility (18045-054). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

CO2 Independent Medium contains no lipids or growth factors. Therefore, CO2 Independent Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). The CO2 Independent Medium does not require a CO2 environment to maintain physiological pH. For maximum growth performance some cell lines may require either direct or sequential adaptation to CO2 Independent Medium (see protocol).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineEpithelial, fibroblast, and lymphoid
Concentration1 X
Product TypeCO2 Independent Medium
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
SpeciesMouse Embryos
FormLiquid
SterilitySterile-filtered
With AdditivesPhenol Red, Sodium Pyruvate
Without AdditivesNo Glutamine, No HEPES
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.