Allyl acetate, 98%
Allyl acetate, 98%
Allyl acetate, 98%
Thermo Scientific Chemicals

Allyl acetate, 98%

CAS: 591-87-7 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 180655000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS591-87-7
IUPAC Nameprop-2-en-1-yl acetate
Molecular FormulaC5H8O2
InChI KeyFWZUNOYOVVKUNF-UHFFFAOYSA-N
SMILESCC(=O)OCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4030 to 1.4090 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only