DNase I, Amplification Grade
Invitrogen™

DNase I, Amplification Grade

DNase I, Amplification Grade, digests single- and double-stranded DNA to oligodexyribonuleotides containing a 5' phosphate. DNase I, Amplification Grade, isRead more
Have Questions?
Catalog NumberQuantity
18068015100 U
Catalog number 18068015
Price (USD)
210.00
Each
Quantity:
100 U
Request bulk or custom format
Price (USD)
210.00
Each
DNase I, Amplification Grade, digests single- and double-stranded DNA to oligodexyribonuleotides containing a 5' phosphate. DNase I, Amplification Grade, is suitable for eliminating DNA during critical RNA purification procedures such as those prior to RNA-PCR amplification. It is purified and tested for non-detectable levels of RNase contamination. Absence of RNase is tested by performing a ribonuclease assay with RNA ladder.

Application
Removing DNA from RNA and protein preparations.

Specific activity
Specific activity is >10,000 units/mg.

Source
Purified from bovine pancreas

Performance and quality testing
Ribonuclease assay with RNA ladder and ability to digest single-stranded and double-stranded DNA to oligonucleotides are determined.

Unit definition
One unit increases the absorbance of a high molecular weight DNA solution at a rate of 0.001 A260 units/min/mL of reaction mixture at 25°C.

Unit reaction conditions
0.1 M sodium acetate (pH 5.0), 5 mM MgCl2, 50 μg/mL calf thymus DNA, and enzyme in 1 mL for 10 min at 25°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferReaction Buffer
Quantity100 U
Shipping ConditionApproved for shipment on Wet or Dry Ice
EnzymeDNase
Unit SizeEach
Contents & Storage
Contents:
1 vial of DNase I, Amp Grade (100 U)
1 vial of 10X DNase I Reaction Buffer (1000 μL)
1 vial of 25 mM EDTA (pH 8.0) (200 μL)

Store at -20°C in a non-frost-free freezer.

Frequently asked questions (FAQs)

How can RNA be treated to remove residual DNA?

RNAse-free DNase treatment of the RNA can reduce DNA to undetectable levels. We recommend using our DNase 1, Amplification Grade (Cat. No. 18068015).

Does Thermo Fisher Scientific offer a protease-free DNase?

We do not test for protease activity as part of our QC but there is PMSF (a protease inhibitor) in the storage buffer. Furthermore, in the preparation of DNase I, we use a soybean trypsin inhibitor column to remove proteases.

How should I treat my RNA sample prior to RT-PCR to ensure that I have no DNA contamination?

DNase I treatment is optional, and one has to consider individual experimental design.

Potential disadvantage of omitting the DNase I step: you may get amplification from genomic DNA. If you omit this step, you will need to include a no RT control and design primers that will not amplify genomic DNA, like those spanning two different exons or exon-exon junctions.

Potential benefit of omitting the DNAse I Step:
saves time; consumes less reagent, saves pipetting steps, and reduces RNA loss (important for precious samples).

Protocol for DNAse I treatment:
Combine 1 µg total RNA, 1 µL 10X DNAse I buffer (200 mM Tris-HCl (pH 8.4), 500 mM KCl, 20 mM MgCl2), 1 µL Amplification Grade DNAse I (1 unit/µL), and DEPC-treated water to 10 µL. Incubate for 15 min at room temperature. Inactivate by adding 1 µL of 25 mM EDTA and heat for 10 min at 65 degrees C.
Note: 1 U of DNAse I should be enough to treat up to ~10 µg of RNA.

To locate the manual for Amplification Grade DNAse I, search www.thermofisher.com with the Cat. No.18068-015. The manual will be one of the links on the product page.

What is the difference between DNase I and Amplification Grade DNase I?

The Amplification Grade DNase I (Cat. No. 18068-015) is subjected to an extra final HPLC purification step to remove traces of RNases. The Amplification Grade DNase I is supplied as 1 unit/µL and comes with 10X buffer (200 mM Tris-HCL pH 8.4, 20 mM MgCl2, 500 mM KCl) and a vial of 25mM EDTA.

In RT-PCR, a large excess of Amplification Grade DNase I could be used to digest an RNA template without degradation of the RNA (in-house data). Use Amplification Grade DNase I to remove genomic DNA carryover in RNA samples prior to RT-PCR.

The regular DNase I is supplied at 5-15 mg/mL (50-375 U/µL) and does not come with its own buffer.

How can I inactivate DNase I?

Add of 1 µL of 25 mM EDTA solution to the reaction mixture in 10 µL reaction with 1 unit DNase I, Amplification Grade (or 1:1 molar ratio of Mg++ ions:EDTA) to chelate the Mg++ ions in the DNase I buffer. Heat for 10 min at 65 degrees C.

Note: It is vital that the EDTA be at least at 2 mM prior to heat-inactivation to avoid Mg-dependent RNA hydrolysis.

DNA-free and Turbo-free versions of DNase I can be inactivated with included DNase Inactivation Reagent.

Citations & References (15)

Citations & References
Abstract
Mapping of multidrug resistance gene 1 and multidrug resistance-associated protein isoform 1 to 5 mRNA expression along the human intestinal tract.
Authors:Zimmermann C,Gutmann H,Hruz P,Gutzwiller JP,Beglinger C,Drewe J
Journal:Drug metabolism and disposition: the biological fate of chemicals
PubMed ID:15523049
Gata factor translation is the final downstream step in the amino acid/tor-mediated vitellogenin gene expression in the anautogenous mosquito aedes aegypti.
Authors:Park JH, Attardo GM, Hansen IA, Raikhel AS,
Journal:J Biol Chem
PubMed ID:16490782
'Ingestion of blood is required for vector mosquitoes to initiate reproductive cycles determining their role as vectors of devastating human diseases. Nutritional signaling plays a pivotal role in regulating mosquito reproduction. Transcription of yolk protein precursor genes is repressed until mosquitoes take blood. Previously, we have shown that to signal ... More
Transcriptional effects of chronic Akt activation in the heart.
Authors: Cook Stuart A; Matsui Takashi; Li Ling; Rosenzweig Anthony;
Journal:J Biol Chem
PubMed ID:11956204
'Akt activation reduces cardiomyocyte death and induces cardiac hypertrophy. To help identify effector mechanisms, gene expression profiles in hearts from transgenic mice with cardiac-specific expression of activated Akt (myr-Akt) were compared with littermate controls. 40 genes were identified as differentially expressed. Quantitative reverse transcription-PCR confirmed qualitative results of transcript profiling ... More
CCAAT/Enhancer Binding Protein alpha Binds to the Epstein-Barr Virus (EBV) ZTA Protein through Oligomeric Interactions and Contributes to Cooperative Transcriptional Activation of the ZTA Promoter through Direct Binding to the ZII and ZIIIB Motifs during Induction of the EBV Lytic Cycle.
Authors:Wu FY, Wang SE, Chen H, Wang L, Hayward SD, Hayward GS,
Journal:J Virol
PubMed ID:15078966
'The Epstein-Barr virus (EBV)-encoded ZTA protein interacts strongly with and stabilizes the cellular CCAAT/enhancer binding protein alpha (C/EBPalpha), leading to the induction of p21-mediated G(1) cell cycle arrest. Despite the strong interaction between these two basic leucine zipper (bZIP) family proteins, the ZTA and C/EBPalpha subunits do not heterodimerize, as ... More
Transcriptional Regulation of the Pituitary Vasopressin V1b Receptor Involves a GAGA-binding Protein.
Authors: Volpi Simona; Rabadan-Diehl Cristina; Cawley Niamh; Aguilera Greti;
Journal:J Biol Chem
PubMed ID:12023277
'The role of CT repeats (inverted GAGA box) in the rat vasopressin V1b receptor (V1bR) promoter in the transcriptional regulation of this gene was studied in H32 hypothalamic cells, which express endogenous V1bR. Transfection of a 2.5-kb V1bR fragment (2161 bp upstream and 377 bp downstream of the proximal transcriptional ... More