Terephthalic acid, 99+%
Terephthalic acid, 99+%
Terephthalic acid, 99+%
Thermo Scientific Chemicals

Terephthalic acid, 99+%

CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 180720250
Price (USD)
-
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Chemical Identifiers
CAS100-21-0
IUPAC Namebenzene-1,4-dicarboxylic acid
Molecular FormulaC8H6O4
InChI KeyKKEYFWRCBNTPAC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.2 %
Titration with TBAH>=99.0 %
Sulfated ash=<0.001 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only