1-Tetradecanol, 99%
1-Tetradecanol, 99%
1-Tetradecanol, 99%
Thermo Scientific Chemicals

1-Tetradecanol, 99%

CAS: 112-72-1 | C14H30O | 214.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 180730250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-72-1
IUPAC Nametetradecan-1-ol
Molecular FormulaC14H30O
InChI KeyHLZKNKRTKFSKGZ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes or
Appearance (Form)pellets and/or chunks
Infrared spectrumConforms
Melting point37°C to 41°C
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only