1-Tetradecanol, 99%
1-Tetradecanol, 99%
1-Tetradecanol, 99%
Thermo Scientific Chemicals

1-Tetradecanol, 99%

CAS: 112-72-1 | C14H30O | 214.39 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 180731000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS112-72-1
IUPAC Nametetradecan-1-ol
Molecular FormulaC14H30O
InChI KeyHLZKNKRTKFSKGZ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCO
さらに表示
Appearance (Form)Crystalline powder or crystals or flakes or
Appearance (Form)pellets and/or chunks
Infrared spectrumConforms
Melting point37°C to 41°C
Appearance (Color)White
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only