Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Thermo Scientific Chemicals

Phenol, 99+%, for biochemistry, loose crystals

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 180781000
Price (USD)
103.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
103.00
Each
Chemical Identifiers
CAS108-95-2
IUPAC Namephenol
Molecular FormulaC6H6O
InChI KeyISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Loose crystals or flakes
GC>=99.0 %
Water=<0.5 %
Residue after evaporation=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only