2-Naphthol, 99+%
2-Naphthol, 99+%
2-Naphthol, 99+%
Thermo Scientific Chemicals

2-Naphthol, 99+%

CAS: 135-19-3 | C10H8O | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 180821000
Price (USD)
44.20
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.20
Each
Chemical Identifiers
CAS135-19-3
IUPAC Namenaphthalen-2-ol
Molecular FormulaC10H8O
InChI KeyJWAZRIHNYRIHIV-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)White to beige to pink to light brown
Appearance (Form)Powder or flakes
Infrared spectrumConforms
Melting point120°C to 124°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only