Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Thermo Scientific Chemicals

Propionitrile, 99%, extra pure

CAS: 107-12-0 | C3H5N | 55.08 g/mol
数量:
100 mL
500 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 180895000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS107-12-0
IUPAC Namepropanenitrile
Molecular FormulaC3H5N
InChI KeyFVSKHRXBFJPNKK-UHFFFAOYSA-N
SMILESCCC#N
さらに表示
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3650 to 1.3680 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only