Naphthalene, 99%
Naphthalene, 99%
Naphthalene, 99%
Thermo Scientific Chemicals

Naphthalene, 99%

CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 180902500
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.20
Save 4.55
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.20
Save 4.55
Each
Chemical Identifiers
CAS91-20-3
IUPAC Namenaphthalene
Molecular FormulaC10H8
InChI KeyUFWIBTONFRDIAS-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)tablets or balls
Melting point79°C to 82°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only