Product Image
Thermo Scientific Chemicals

Benzyl cyanide, 99+%

CAS: 140-29-4 | C8H7N | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 180940010
Price (TWD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS140-29-4
IUPAC Name2-phenylacetonitrile
Molecular FormulaC8H7N
InChI KeySUSQOBVLVYHIEX-UHFFFAOYSA-N
SMILESN#CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5220 to 1.5240 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only