4-chlorophenol, 99+%
4-chlorophenol, 99+%
4-chlorophenol, 99+%
Thermo Scientific Chemicals

4-chlorophenol, 99+%

CAS: 106-48-9 | C6H5ClO | 128.56 g/mol
数量:
100 g
500 g
5 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 181000051
価格(JPY)
-
数量:
5 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS106-48-9
IUPAC Name4-chlorophenol
Molecular FormulaC6H5ClO
InChI KeyWXNZTHHGJRFXKQ-UHFFFAOYSA-N
SMILESOC1=CC=C(Cl)C=C1
さらに表示
Appearance (Color)White to light brown
Appearance (Form)Crystals or fused mass
Infrared spectrumConforms
GC>=99.0 %
Solubility(5 % in ethanol) Clear to slightly hazy colorless to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only