Ethyl chloroformate, 97%
Ethyl chloroformate, 97%
Ethyl chloroformate, 97%
Thermo Scientific Chemicals

Ethyl chloroformate, 97%

CAS: 541-41-3 | C3H5ClO2 | 108.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 181080050
Price (EUR)
35,70
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
35,70
Each
Chemical Identifiers
CAS541-41-3
IUPAC Nameethyl carbonochloridate
Molecular FormulaC3H5ClO2
InChI KeyRIFGWPKJUGCATF-UHFFFAOYSA-N
SMILESCCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<100 APHA
GC>=96.0 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.3940 to 1.3960 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only