2-Furaldehyde, 99%
2-Furaldehyde, 99%
2-Furaldehyde, 99%
Thermo Scientific Chemicals

2-Furaldehyde, 99%

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 181100250
Price (EUR)
28,70
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
28,70
Each
Chemical Identifiers
CAS98-01-1
IUPAC Namefuran-2-carbaldehyde
Molecular FormulaC5H4O2
InChI KeyHYBBIBNJHNGZAN-UHFFFAOYSA-N
SMILESO=CC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear amber to brown
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Water=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only