Furan, 99+%, stabilized
Furan, 99+%, stabilized
Furan, 99+%, stabilized
Thermo Scientific Chemicals

Furan, 99+%, stabilized

CAS: 110-00-9 | C4H4O | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 181112500
Price (EUR)
31,65
Online exclusive
35,10
Save 3,45 (10%)
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
31,65
Online exclusive
35,10
Save 3,45 (10%)
Each
Chemical Identifiers
CAS110-00-9
IUPAC Namefuran
Molecular FormulaC4H4O
InChI KeyYLQBMQCUIZJEEH-UHFFFAOYSA-N
SMILESO1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.2 %
Stabilizer0.025 to 0.04 % BHT
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only