3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
Thermo Scientific Chemicals

3,5-Dimethylpiperidine, 96%, mixture of cis and trans

CAS: 35794-11-7 | C7H16N | 114.21 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 181230250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS35794-11-7
IUPAC Name(3R,5S)-3,5-dimethylpiperidin-1-ium
Molecular FormulaC7H16N
InChI KeyIDWRJRPUIXRFRX-KNVOCYPGSA-O
SMILESC[C@H]1C[NH2+]C[C@@H](C)C1
さらに表示
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=95 % (sum of cis and trans)
Refractive index1.4425 to 1.4475 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only