3-Bromobenzoic acid, 99%
3-Bromobenzoic acid, 99%
3-Bromobenzoic acid, 99%
Thermo Scientific Chemicals

3-Bromobenzoic acid, 99%

CAS: 585-76-2 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
181351000100 gPlastic bottle
Catalog number 181351000
Price (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS585-76-2
IUPAC Name3-bromobenzoic acid
Molecular FormulaC7H5BrO2
InChI KeyVOIZNVUXCQLQHS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point154°C to 160°C
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only