N,N-Diethylformamide, 99%
N,N-Diethylformamide, 99%
N,N-Diethylformamide, 99%
Thermo Scientific Chemicals

N,N-Diethylformamide, 99%

CAS: 617-84-5 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 181370250
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.60
Save 5.95
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.60
Save 5.95
Each
Chemical Identifiers
CAS617-84-5
IUPAC NameN,N-diethylformamide
Molecular FormulaC5H11NO
InChI KeySUAKHGWARZSWIH-UHFFFAOYSA-N
SMILESCCN(CC)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only