(S)-(+)-2-Amino-3-methyl-1-butanol, 96%
(S)-(+)-2-Amino-3-methyl-1-butanol, 96%
(S)-(+)-2-Amino-3-methyl-1-butanol, 96%
Thermo Scientific Chemicals

(S)-(+)-2-Amino-3-methyl-1-butanol, 96%

CAS: 2026-48-4 | C5H13NO | 103.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 181590250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2026-48-4
IUPAC Name(2S)-2-amino-3-methylbutan-1-ol
Molecular FormulaC5H13NO
InChI KeyNWYYWIJOWOLJNR-RXMQYKEDSA-N
SMILESCC(C)[C@H](N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting or solid
GC>=95 %
Specific optical rotation+15° to +19° (20°C, 589 nm) (c=10, ethanol)
Refractive index1.4535 to 1.4565 (20°C, 589 nm)
Appearance (Color)Clear colorless to white to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only