2-Bromobenzyl bromide, 98%
2-Bromobenzyl bromide, 98%
2-Bromobenzyl bromide, 98%
Thermo Scientific Chemicals

2-Bromobenzyl bromide, 98%

CAS: 3433-80-5 | C7H6Br2 | 249.93 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 181790250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3433-80-5
IUPAC Name1-bromo-2-(bromomethyl)benzene
Molecular FormulaC7H6Br2
InChI KeyLZSYGJNFCREHMD-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1Br
さらに表示
Appearance (Form)Liquid after melting
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.6170 to 1.6190 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only