Cinnamyl chloride, 95% trans
Cinnamyl chloride, 95% trans
Cinnamyl chloride, 95% trans
Thermo Scientific Chemicals

Cinnamyl chloride, 95% trans

CAS: 2687-12-9 | C9H9Cl | 152.62 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
產品號碼 181850050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2687-12-9
IUPAC Name[(1E)-3-chloroprop-1-en-1-yl]benzene
Molecular FormulaC9H9Cl
InChI KeyIWTYTFSSTWXZFU-QPJJXVBHSA-N
SMILESClC\C=C\C1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
GC>=94 %
Appearance (Color)Colorless to yellow to yellow-brown
Infrared spectrumConforms
Refractive index1.5830 to 1.5850 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only