2,6-Dichlorobenzoyl chloride, 98%
2,6-Dichlorobenzoyl chloride, 98%
2,6-Dichlorobenzoyl chloride, 98%
Thermo Scientific Chemicals

2,6-Dichlorobenzoyl chloride, 98%

CAS: 4659-45-4 | C7H3Cl3O | 209.46 g/mol
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 182060500
Price (JPY)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4659-45-4
IUPAC Name2,6-dichlorobenzoyl chloride
Molecular FormulaC7H3Cl3O
InChI KeyJBLIDPPHFGWTKU-UHFFFAOYSA-N
SMILESClC(=O)C1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration RCOCl97.5 to 102.5 %
Refractive index1.5590 to 1.5610 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow to light pink
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only