3-Methoxyphenethyl alcohol, 97%
3-Methoxyphenethyl alcohol, 97%
3-Methoxyphenethyl alcohol, 97%
Thermo Scientific Chemicals

3-Methoxyphenethyl alcohol, 97%

CAS: 5020-41-7 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 182250050
Price (EUR)
164,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
164,00
Each
Chemical Identifiers
CAS5020-41-7
IUPAC Name2-(3-methoxyphenyl)ethan-1-ol
Molecular FormulaC9H12O2
InChI KeyUPPGEJSCUZMCMW-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CCO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only