o-Cresolphthalein Complexone, pure
o-Cresolphthalein Complexone, pure
o-Cresolphthalein Complexone, pure
Thermo Scientific Chemicals

o-Cresolphthalein Complexone, pure

CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 182440250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2411-89-4
IUPAC Name2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
Molecular FormulaC32H32N2O12
InChI KeyIYZPEGVSBUNMBE-UHFFFAOYSA-N
SMILESCC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Suitability(as complexom. indicator) Passes test
Appearance (Form)Powder
UV Lambda max573 to 579 nm (in 0.1N NaOH)
Appearance (Color)Off-white to pink-beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only