2-Hydroxyphenethyl alcohol, 98%
2-Hydroxyphenethyl alcohol, 98%
2-Hydroxyphenethyl alcohol, 98%
Thermo Scientific Chemicals

2-Hydroxyphenethyl alcohol, 98%

CAS: 7768-28-7 | C8H10O2 | 138.17 g/mol
製品番号(カタログ番号)数量パッケージ
1824900101 gGlass bottle
製品番号(カタログ番号) 182490010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7768-28-7
IUPAC Name2-(2-hydroxyethyl)phenol
Molecular FormulaC8H10O2
InChI KeyABFCOJLLBHXNOU-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1O
さらに表示
Appearance (Form)Viscous liquid
Refractive index1.5560 to 1.5600 (20°C, 589 nm)
Appearance (Color)Clear yellow to yellow-brown
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only