4-Hydroxyphenethyl alcohol, 98%
4-Hydroxyphenethyl alcohol, 98%
4-Hydroxyphenethyl alcohol, 98%
Thermo Scientific Chemicals

4-Hydroxyphenethyl alcohol, 98%

CAS: 501-94-0 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 182500250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS501-94-0
IUPAC Name4-(2-hydroxyethyl)phenol
Molecular FormulaC8H10O2
InChI KeyYCCILVSKPBXVIP-UHFFFAOYSA-N
SMILESOCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Off-white to light beige
Appearance (Form)Crystalline powder or crystals
Melting point89°C to 93°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only