This product ships via ground from US northeast.
D(+)-Trehalose dihydrate, 99%
D(+)-Trehalose dihydrate, 99%
D(+)-Trehalose dihydrate, 99%
Thermo Scientific Chemicals

D(+)-Trehalose dihydrate, 99%

CAS: 6138-23-4 | C12H26O13 | 378.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 182555000
Price (USD)
916.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,078.00
Save 161.35 (15%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
916.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,078.00
Save 161.35 (15%)
Each
Chemical Identifiers
CAS6138-23-4
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
Molecular FormulaC12H26O13
InChI KeyDPVHGFAJLZWDOC-DJCWUSJTNA-N
SMILESO.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Water8.5 to 10.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only