Library Efficiency™ DH5α Competent Cells
Invitrogen™
Library Efficiency™ DH5α Competent Cells
Library Efficiency DH5α Competent Cells are a versatile strain of chemically competent cells that are used in many everyday cloningRead more
Have Questions?
Catalog NumberQuantity
182630125 x 200 μL
Catalog number 18263012
Price (USD)
289.00
Each
Quantity:
5 x 200 μL
Price (USD)
289.00
Each
Library Efficiency DH5α Competent Cells are a versatile strain of chemically competent cells that are used in many everyday cloning applications. Prepared by a proprietary modification of the Hanahan method, these cells are able to achieve transformation efficiencies of >1 x 108 cfu/μg control DNA per 100 μL reaction.  
Library Efficiency DH5α Competent Cells are recommended for routine cloning of genes into large plasmids and are suitable for difficult clone constructions, such as blunt-end ligations or cloning using limited amount of DNA.
The strain has the lacZΔM15 genotype, allowing blue-white screening on plates containing either X-Gal or Bluo-Gal. It also carries the recA1 mutation, which helps reduce the rate of recombination while propagating plasmid DNA, and the endA1 mutation that makes DH5α cells an excellent choice for amplifying plasmid DNA for subsequent extraction and purification. DH5α cells are capable of being transformed efficiently with large plasmids and can serve as a host for the M13mp cloning vectors if a lawn of DH5αF′IQ, for example, is provided to allow plaque formation.
Library Efficiency DH5α Competent Cells offer: 
• Transformation efficiencies of >1 x 108 cfu/μg
• lacZΔM15 marker that provides α-complementation of the β-galactosidase gene allowing blue/white screening on agar plates containing X-Gal or Bluo-Gal
• Plasmid DNA maintained with minimum recombination events due to recA1 mutation
• endA1 mutation that enables increased plasmid yield and quantity
• hsdR17 genotype that allows efficient transformation of cloned unmethylated DNA from PCR
Genotype
F- Φ80lacZΔM15 Δ(lacZYA-argF) U169 recA1 endA1 hsdR17 (rk-, mk+) phoA supE44 λ-thi-1 gyrA96 relA1
Find the strain and format that fits your needs 
DH5α strain and its derivatives are available with a variety of transformation efficiencies and in both electrocompetent and chemically competent formats.
MAX Efficiency DH5αF´IQ Competent Cells are available in chemically competent format.
DH5α-T1R competent cells are available in MultiShot format for high throughput applications.
Explore bacterial growth media formats.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialNo
Blue/White ScreeningYes (lacZΔM15)
Cloning Methylated DNANo
Cloning Unstable DNANot suitable for cloning unstable DNA
Contains F' EpisomeNo
High-throughput CompatibilityLow
Improves Plasmid QualityYes (endA1)
Preparing Unmethylated DNANo
Product LineLibrary Efficiency
Product TypeChemically Competent Cells
Quantity5 x 200 μL
Reduces RecombinationYes (recA1)
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)No
Transformation Efficiency LevelMedium Efficiency (1 x 108 to 1 x 109 cfu/μg)
FormatTube
SpeciesE. coli (K12)
Unit SizeEach
Contents & Storage
• Library Efficiency DH5α Competent Cells (5 x 200 μL)
Store Competent Cells at –80°C.
• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.
• S.O.C. Medium (2 x 6 mL)
Store S.O.C. Medium at 4°C or room temperature.
Frequently asked questions (FAQs)
I am trying to clone an insert that is supposedly pretty toxic. I used DH5? and TOP10 cells for the transformation and got no colonies on the plate. Do you have any suggestions for me?
If the insert is potentially toxic to the host cells, here are some suggestions that you can try:
- After transforming TOP10 or DH5? cells, incubate at 25-30°C instead of 37°C. This will slow down the growth and will increase the chances of cloning a potentially toxic insert.
- Try using TOP10F' cells for the transformation, but do not add IPTG to the plates. These cells carry the lacIq repressor that represses expression from the lac promoter and so allows cloning of toxic genes. Keep in mind that in the absence of IPTG, blue-white screening cannot be performed.
- Try using Stbl2 cells for the transformation.
How do you recommend that I prepare my DNA for successful electroporation of E. coli?
For best results, DNA used in electroporation must have a very low ionic strength and a high resistance. A high-salt DNA sample may be purified by either ethanol precipitation or dialysis.
The following suggested protocols are for ligation reactions of 20ul. The volumes may be adjusted to suit the amount being prepared.
Purifying DNA by Precipitation: Add 5 to 10 ug of tRNA to a 20ul ligation reaction. Adjust the solution to 2.5 M in ammonium acetate using a 7.5 M ammonium acetate stock solution. Mix well. Add two volumes of 100 % ethanol. Centrifuge at 12,000 x g for 15 min at 4C. Remove the supernatant with a micropipet. Wash the pellet with 60ul of 70% ethanol. Centrifuge at 12,000 x g for 15 min at room temperature. Remove the supernatant with a micropipet. Air dry the pellet. Resuspend the DNA in 0.5X TE buffer [5 mM Tris-HCl, 0.5 mM EDTA (pH 7.5)] to a concentration of 10 ng/ul of DNA. Use 1 ul per transformation of 20 ul of cell suspension.
Purifying DNA by Microdialysis: Float a Millipore filter, type VS 0.025 um, on a pool of 0.5X TE buffer (or 10% glycerol) in a small plastic container. Place 20ul of the DNA solution as a drop on top of the filter. Incubate at room temperature for several hours. Withdraw the DNA drop from the filter and place it in a polypropylene microcentrifuge tube. Use 1ul of this DNA for each electrotransformation reaction.
You offer competent cells in Subcloning Efficiency, Library Efficiency and MAX Efficiency. How do these differ?
There are a few exceptions, but in general the difference is in guaranteed transformation efficiency as follows:
Subcloning Efficiency cells are guaranteed to produce at least 1.0 x 10E6 transformants per µg of transformed pUC19 or pUC18 supercoiled plasmid
Library Efficiency cells are guaranteed to produce at least 1.0 x 10E8 transformants per µg pUC19 or pUC18 DNA
MAX Efficiency cells are guaranteed to produce at least 1.0 x 10E9 transformants per µg pUC19 or pUC18 DNA
Do any Invitrogen competent cells contain DMSO in the freezing medium?
Yes, several of our competent cells products are frozen with DMSO. The presence of DMSO (dimethylsulfoxide) will generally be indicated in the MSDS files if you have a question about a particular product, but here is a list of commonly used products that are known to have DMSO in the freezing buffer:
One Shot OmniMAX 2 T1 Phage Resistant Cells, Cat. No. C8540-03
One Shot INV?F' Chemically Competent Cells, Cat. No. C2020-03 and C2020-06
One Shot MAX Efficiency DH5?-T1 Chemically Competent Cells, Cat. No. 12297-016
MAX Efficiency DH5?-T1 Phage Resistant Cells, Cat. No. 12034-013
MAX Efficiency DH5? Chemically Competent Cells, Cat. No. 18258-012
Library Efficiency DH5? Chemically Competent Cells, Cat. No. 18263-012
MAX Efficiency DH5? F'IQ Cells, Cat. No. 18288-019
MAX Efficiency Stbl2Chemically Competent Cells, Cat. No. 10268-019
Is S.O.C. medium absolutely required when recovering competent bacterial cells during transformation?
Many media can be used to grow transformed cells, including standard LB, SOB or TB broths. However, S.O.C. is the optimal choice for recovery of the cells before plating. The nutrient-rich formula with added glucose is often important for obtaining maximum transformation efficiencies.
Citations & References (4)
Citations & References
Abstract
Changes in the mechanism of DNA integration in vitro induced by base substitutions in the HIV-1 U5 and U3 terminal sequences.
Authors: Brin Elena; Leis Jonathan;
Journal:J Biol Chem
PubMed ID:11788585
'We have reconstituted concerted human immunodeficiency virus type 1 (HIV-1) integration with specially designed mini-donor DNA, a supercoiled plasmid acceptor, purified bacterial-derived HIV-1 integrase (IN), and host HMG-I(Y) protein (Hindmarsh, P., Ridky, T., Reeves, R., Andrake, M., Skalka, A. M., and Leis, J. (1999) J. Virol. 73, 2994-3003). Integration in ... More
Identification and characterization of two cation binding sites in the integrin beta 3 subunit.
Authors: Cierniewska-Cieslak Aleksandra; Cierniewski Czeslaw S; Blecka Kamila; Papierak Malgorzata; Michalec Lidia; Zhang Li; Haas Thomas A; Plow Edward F;
Journal:J Biol Chem
PubMed ID:11796735
'The midsegment of the beta(3) subunit has been implicated in the ligand and cation binding functions of the beta(3) integrins. This region may contain a metal ion-dependent adhesion site (MIDAS) and fold into an I domain-like structure. Two recombinant fragments, beta(3)-(95-373) and beta(3)-(95-301), were expressed and found to bind fibrinogen. ... More
Requirement for either a host- or pectin-induced pectate lyase for infection of pisum sativum by nectriahematococca.
Authors:Rogers LM, Kim YK, Guo W, Gonzalez-Candelas L, Li D, Kolattukudy PE
Journal:Proc Natl Acad Sci U S A
PubMed ID:10931947
Fungal pathogens usually have multiple genes that encode extracellular hydrolytic enzymes that may degrade the physical barriers in their hosts during the invasion process. Nectria hematococca, a plant pathogen, has two inducible pectate lyase (PL) genes (pel) encoding PL that can help degrade the carbohydrate barrier in the host. pelA ... More
Molecular determinants of high affinity binding to group III metabotropic glutamate receptors.
Authors: Rosemond Erica; Peltekova Vanya; Naples Mark; Thøgersen Henning; Hampson David R;
Journal:J Biol Chem
PubMed ID:11744707
The amino-terminal domain containing the ligand binding site of the G protein-coupled metabotropic glutamate receptors (mGluRs) consists of two lobes that close upon agonist binding. In this study, we explored the ligand binding pocket of the Group III mGluR4 receptor subtype using site-directed mutagenesis and radioligand binding. The selection of ... More