Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
Bottle Surface | Smooth |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 20 |
Packaging | 20/Pk., 20/Case; Double-bagged |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Closure Included | Yes |
Closure Type | Easy on/off, Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Closure Included | Yes |
Closure Type | Easy on/off, Vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 20 |
Packaging | 20/Pk., 20/Case |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Closure Included | Yes |
Closure Type | Easy on/off, Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Closure Included | Yes |
Closure Type | Easy on/off, Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth surface, shallow indent |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Closure Included | Yes |
Closure Type | EZ Twist Vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth surface, deep indent |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Graduations | Yes |
Unit Size | Case of 20 |
Bottle Surface | Smooth surface, shallow indent |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Graduations | Yes |
Closure Included | Yes |
Closure Material | HDPE |
Closure Type | EZ Twist Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Mouth | Narrow |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth surface, shallow indent |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 20 |
Packaging | 20/Pk., 20/Case; Double-bagged |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Graduations | Yes |
Closure Included | Yes |
Closure Material | HDPE |
Closure Type | EZ Twist Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Mouth | Narrow |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth surface, deep indent |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 2 |
Packaging | 2/Pk., 20/Case; Double-bagged |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Graduations | Yes |
Closure Included | Yes |
Closure Material | HDPE |
Closure Type | EZ Twist Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Mouth | Narrow |
Shape | Round |
Unit Size | Case of 20 |
Bottle Surface | Smooth surface, deep indent |
Color | Clear |
Material | Polystyrene |
No. per Case | 20 |
No. per Pack | 20 |
Packaging | 20/Pk., 20/Case |
Sterility | Sterile |
Type | Roller Bottle |
Cell Growth Area | 850 cm2 |
Graduations | Yes |
Closure Included | Yes |
Closure Material | HDPE |
Closure Type | EZ Twist Non-vented Cap |
For Use With (Equipment) | Cell Culture Expansion, Vaccine Production, Antibody Production |
Graduated | Yes |
Mouth | Narrow |
Shape | Round |
Unit Size | Case of 20 |
Catalog Number | Specifications | Unit Size | Cell Growth Area | Bottle Surface | Packaging | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
182744 | Case of 20 | 850 cm2 | Smooth | 20/Pk., 20/Case; Double-bagged | 189.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
181702 | Case of 20 | 850 cm2 | Smooth | 2/Pk., 20/Case | 249.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
182720 | Case of 20 | 850 cm2 | Smooth | 20/Pk., 20/Case | 171.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
182702 | Case of 20 | 850 cm2 | Smooth | 2/Pk., 20/Case | 209.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
183302 | Case of 20 | 850 cm2 | Smooth surface, shallow indent | 2/Pk., 20/Case | 265.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
183902 | Case of 20 | 850 cm2 | Smooth surface, deep indent | 2/Pk., 20/Case | 268.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
184302 | Case of 20 | 850 cm2 | Smooth surface, shallow indent | 2/Pk., 20/Case | 228.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
184344 | Case of 20 | 850 cm2 | Smooth surface, shallow indent | 20/Pk., 20/Case; Double-bagged | 206.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
184902 | Case of 20 | 850 cm2 | Smooth surface, deep indent | 2/Pk., 20/Case; Double-bagged | 225.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
184920 | Case of 20 | 850 cm2 | Smooth surface, deep indent | 20/Pk., 20/Case | 186.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|