15-Crown-5, 98%
15-Crown-5, 98%
15-Crown-5, 98%
Thermo Scientific Chemicals

15-Crown-5, 98%

CAS: 33100-27-5 | C10H20O5 | 220.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 182860050
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.20
Save 9.55
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.20
Save 9.55
Each
Chemical Identifiers
CAS33100-27-5
IUPAC Name1,4,7,10,13-pentaoxacyclopentadecane
Molecular FormulaC10H20O5
InChI KeyVFTFKUDGYRBSAL-UHFFFAOYSA-N
SMILESC1COCCOCCOCCOCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4640 to 1.4670 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only