Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™
Agarose (4%)
Catalog number: 18300012
Have Questions?
Gibco™
Agarose (4%)
Catalog number: 18300012
Have Questions?
Catalog Number
18300012
Unit Size
40 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1830001240 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ 4% Agarose is an autoclaved, high purity, low melting point gel designed for plaque assays and the purification of baculovirus. The 4% Agarose Gel melts at 65 – 70°C and remains fluid at 37°C. When diluted with concentrated insect cell culture medium, Gibco™ 4% Agarose produces a gel firm enough to immobilize local infections, yet ductile enough to allow for the efficient removal of plaques for clonal expansion.
Product Use
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell Type
Mammalian Cells
Concentration
0.04
Form
Gel
Product Type
Agarose
Shelf Life
12 Months
Sterility
Sterile
Sterilization Method
Autoclaved
For Use With (Application)
Plaque Assay
Remarks
Not for electrophoresis
Shipping Condition
Room Temperature
Contents & Storage
Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Figures
Documents & Downloads
Certificates
    Frequently asked questions (FAQs)
    Citations & References
    Search citations by name, author, journal title or abstract text