ElectroMAX™ A. tumefaciens LBA4404 Cells are Agrobacterium cells that have been specially prepared for transformation by electroporation. ElectroMAX™ LBA4404 CellsLeia mais
Have Questions?
Número do catálogo
Quantity
18313015
5 x 40 μL
Número do catálogo 18313015
Preço (BRL)
5.087,45
Each
Quantity:
5 x 40 μL
Preço (BRL)
5.087,45
Each
ElectroMAX™ A. tumefaciens LBA4404 Cells are Agrobacterium cells that have been specially prepared for transformation by electroporation. ElectroMAX™ LBA4404 Cells contain the disarmed Ti plasmid pAL 4404, which has only the vir and ori region of the Ti plasmid. ElectroMAX™ LBA4404 Cells have been successfully used to transform a variety of plant species, including tobacco and Arabidopsis, at high efficiency.
For Research Use Only. Not for use in diagnostic procedures.
A new class of regulatory genes underlying the cause of pear-shaped tomato fruit.
Authors: Liu Jiping; Van Eck Joyce; Cong Bin; Tanksley Steven D;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12242331
A common, recurring theme in domesticated plants is the occurrence of pear-shaped fruit. A major quantitative trait locus (termed ovate) controlling the transition from round to pear-shaped fruit has been cloned from tomato. OVATE is expressed early in flower and fruit development and encodes a previously uncharacterized, hydrophilic protein with ... More