Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Thermo Scientific Chemicals

Poly(caprolactone) diol, average M.N. 2000

CAS: 36890-68-3 | (C6H10O2)n
数量:
250 g
500 g
2.5 kg
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 183210025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS36890-68-3
Molecular Formula(C6H10O2)n
SMILES*-CCCCCC(=O)O-*
MDL NumberMFCD23713083
Appearance (Color)White
Water=<2 % (K.F.)
Infrared spectrumConforms
Appearance (Form)Waxy solid
Molecular number1500 to 2500
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only