Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Thermo Scientific Chemicals

Poly(caprolactone) diol, average M.N. 2000

CAS: 36890-68-3 | (C6H10O2)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 183212500
Price (EUR)
156,65
Online exclusive
174,00
Save 17,35 (10%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
156,65
Online exclusive
174,00
Save 17,35 (10%)
Each
Chemical Identifiers
CAS36890-68-3
Molecular Formula(C6H10O2)n
SMILES*-CCCCCC(=O)O-*
MDL NumberMFCD23713083
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water=<2 % (K.F.)
Infrared spectrumConforms
Appearance (Form)Waxy solid
Molecular number1500 to 2500
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only