Plumbagin, 99+%
Plumbagin, 99+%
Plumbagin, 99+%
Thermo Scientific Chemicals

Plumbagin, 99+%

CAS: 481-42-5 | C11H8O3 | 188.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1840400101 gGlass bottle
Catalog number 184040010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS481-42-5
IUPAC Name5-hydroxy-2-methyl-1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC11H8O3
InChI KeyVCMMXZQDRFWYSE-UHFFFAOYSA-N
SMILESCC1=CC(=O)C2=C(O)C=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point75°C to 78°C
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)Orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only