4-Methyl-5-thiazoleethanol, 98%
4-Methyl-5-thiazoleethanol, 98%
4-Methyl-5-thiazoleethanol, 98%
Thermo Scientific Chemicals

4-Methyl-5-thiazoleethanol, 98%

CAS: 137-00-8 | C6H9NOS | 143.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 184060250
Price (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS137-00-8
IUPAC Name2-(4-methyl-1,3-thiazol-5-yl)ethan-1-ol
Molecular FormulaC6H9NOS
InChI KeyBKAWJIRCKVUVED-UHFFFAOYSA-N
SMILESCC1=C(CCO)SC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5480 to 1.5510 (20°C, 589 nm)
Appearance (Form)Liquid
Water=<1 %
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only