Oligo(dT)<sub>12-18</sub> Cebador
Invitrogen™

Oligo(dT)12-18 Cebador

El cebador de oligo(dT)12-18 es adecuado para la síntesis del ADNc de primera cadena con transcriptasa inversa.
Have Questions?
Número de catálogoCantidad
1841801250 μL
Número de catálogo 18418012
Precio (USD)
186,62
Each
Cantidad:
50 μL
Precio (USD)
186,62
Each

Oligo(dT)12-18 Primer is suitable for use in first-strand cDNA synthesis with reverse transcriptase. The primer hybridizes to the poly(A) tail of mRNA. It is phosphorylated on the 5' end to facilitate cloning of cDNA.

Performance and quality testing: Performance is evaluated in a first-strand cDNA synthesis reaction

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Modificación del cebador 5'Fosfato
Para utilizar con (aplicación)Síntesis de la primera cadena del ADNc
FormularioLíquido
Secuencia del cebador5'd PO4 [(T)12-18] 3'
Tipo de productoPrimer
Método de purificaciónPurificado de gel, desalado
Cantidad50 μL
Condiciones de envíoAprobado para su envío en hielo húmedo o seco
Concentración50μg/μL
PrimerOligo dT
Unit SizeEach
Contenido y almacenamiento
• Oligo(dT)12-18 Primer in DEPC-treated water (25 μg at 50 μg/μL)

Store at –20°C.
Guaranteed stable for 6 months when properly stored.

Preguntas frecuentes

How does the Anchored Oligo(dT)20 Primer differ from standard oligo(dT) primers?

Anchored Oligo(dT) primers have 2 random bases at the 3' end of the stretch of Ts. The first random base is either an A, G, or C, while the second can be any of the 4 standard nucleotides, i.e., A, T, G or C. The use of anchored oligo(dT) primers results in increased cDNA synthesis yields.

For first-strand cDNA synthesis, is it better to use oligo(dT), random hexamers, gene-specific primer (GSP), or combination of these primers?

The choice of primer depends on your experimental goals. Oligo(dT) is recommended when using total RNA for cDNA synthesis. It is the key to full-length cDNA synthesis. Random hexamers give a series of short first-strand products spanning the entire mRNA. Use of random hexamers may be helpful if the PCR fragment is at the 5´ end of a large mRNA. To ensure full-length cDNA synthesis of large transcripts, oligo(dT) can be added along with random hexamers during first-strand synthesis. Gene-specific primers (GSP) for cDNA synthesis may also be used and are required in a few applications such as 5´ RACE and qRT-PCR. For GC-rich templates or templates rich in secondary structure, a GSP may not work as well as priming with oligo dT for first-strand synthesis. If an RT-PCR is problematic, trying different options of oligo dT, random primers and/or GSP for priming first-strand synthesis may resolve the issue. Oligo(dT)20 primer (Cat. No. 18418-020) is recommended for use with SuperScript III Reverse Transcriptase (Cat. no. 18080-044), ThermoScript Reverse Transcriptase (Cat. No. 12236-014), Thermo-X Reverse Trascriptase (Cat. No. 11150-025), and Cloned AMV Reverse Transcriptase (Cat. No. 12328-019).

Reference:
Frohman,M.A., Dush,M.K., Martin, G.R. (1988) Proc. Nat. Acad. Sci USA 85, 8998

Citations & References (1)

Citations & References
Abstract
The fifth essential DNA polymerase phi in Saccharomyces cerevisiae is localized to the nucleolus and plays an important role in synthesis of rRNA.
Authors: Shimizu Kikuo; Kawasaki Yasuo; Hiraga Shin-Ichiro; Tawaramoto Maki; Nakashima Naomi; Sugino Akio;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12093911
'We report that POL5 encodes the fifth essential DNA polymerase in Saccharomyces cerevisiae. Pol5p was identified and purified from yeast cell extracts and is an aphidicolin-sensitive DNA polymerase that is stimulated by yeast proliferating cell nuclear antigen (PCNA). Thus, we named Pol5p DNA polymerase phi. Temperature-sensitive pol5-1 approximately -3 mutants ... More