Nunc™ Polystyrene Roller Bottles
Nunc™ Polystyrene Roller Bottles
Actual product may vary
Nunc™ Polystyrene Roller Bottles
Nunc™ Polystyrene Roller Bottles
Nunc™ Polystyrene Roller Bottles
Thermo Scientific™
Nunc™ Polystyrene Roller Bottles
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell Growth AreaBottle SurfaceClosure TypePackaging
184344850 cm2Smooth, shallow indentEZ Twist Non-vented Cap20/Pk., 20/Cs. (Double-bagged)
181702850 cm2Smooth, no indentEasy on/off, Vented Cap2/Pk., 20/Cs.
182720850 cm2Smooth, no indentEasy on/off, Non-vented Cap20/Pk., 20/Cs.
182744850 cm2Smooth, no indentEasy on/off, Non-vented Cap20/Pk., 20/Cs. (Double-bagged)
182702850 cm2Smooth, no indentEasy on/off, Non-vented Cap2/Pk., 20/Cs.
183302850 cm2Smooth, shallow indentEZ Twist Vented Cap2/Pk., 20/Cs.
183902850 cm2Smooth, deep indentEZ Twist Vented Cap2/Pk., 20/Cs.
184302850 cm2Smooth, shallow indentEZ Twist Non-vented Cap2/Pk., 20/Cs.
184902850 cm2Smooth, deep indentEZ Twist Non-vented Cap2/Pk., 20/Cs. (Double-bagged)
184920850 cm2Smooth, deep indentEZ Twist Non-vented Cap20/Pk., 20/Cs.
184944850 cm2Smooth, deep indentEZ Twist Non-vented Cap20/Pk., 20/Cs. (Double-bagged)
1427441450 cm2Pleated, no indentEasy on/off, Non-vented Cap20/Pk., 20/Cs. (Double-bagged)
1417441450 cm2Pleated, no indentEasy on/off, Vented Cap20/Pk., 20/Cs. (Double-bagged)
1427201450 cm2Pleated, no indentEasy on/off, Non-vented Cap20/Pk., 20/Cs.
Catalog number 184344
Price (USD)
257.00
Case of 20
Add to cart
Cell Growth Area:
850 cm2
Bottle Surface:
Smooth, shallow indent
Closure Type:
EZ Twist Non-vented Cap
Packaging:
20/Pk., 20/Cs. (Double-bagged)
Price (USD)
257.00
Case of 20
Add to cart
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Features:
  • Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
  • One-piece seamless design reduces possibility of leaking through a seam.
  • Shallow and deep indents allow for excellent manual and automated handling.
  • Proprietary manufacturing process allows for greater impact resistance
  • Vertical expanded surfaces version facilitates emptying and reduces retention of product
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Bottle SurfaceSmooth, shallow indent
Cell Growth Area850 cm2
Closure MaterialHDPE
Closure TypeEZ Twist Non-vented Cap
For Use With (Application)Cell Culture Expansion, Vaccine Production, Antibody Production
No. per Case20
No. per Pack20
Packaging20/Pk., 20/Cs. (Double-bagged)
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPolystyrene
MouthNarrow
Product TypeRoller Bottle
ShapeRound
SterilitySterile
Unit SizeCase of 20