Copper(II) sulfate pentahydrate, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Copper(II) sulfate pentahydrate, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets BP, EP, and USP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
12 kg
Número de catálogo 1846-57
Precio (USD)
-
Cantidad:
100 g
This product is GMP Manufactured and meets BP, EP, and USP specifications.
This product is GMP Manufactured and meets BP, EP, and USP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición150°C (1013 mmHg)
CAS7758-99-8
Nombre del producto químico o materialCopper(II) sulfate pentahydrate
ColorAzul
Punto de fusión110°C
Fórmula molecularCuSO4·5H2O
EnvaseAmber glass bottle, narrow mouth
SinónimoCupric sulfate pentahydrate, Copper sulfate pentahydrate, Copper sulphate pentahydrate, Cupric sulphate pentahydrate
GradoMulticomponente/USP
Peso molecular (g/mol)249.69
Porcentaje de pureza98.5 to 100.5%
Forma físicaSolid
Cantidad100 g
pH3.5 to 4.5 (50 g/L, H2O, 20°C)
Unit Size100 g