Glycerol, 99.5+%, for spectroscopy
Glycerol, 99.5+%, for spectroscopy
Glycerol, 99.5+%, for spectroscopy
Thermo Scientific Chemicals

Glycerol, 99.5+%, for spectroscopy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
184695000500 mLGlass Bottle
Catalog number 184695000
Price (USD)
153.00
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
153.00
Each
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
UV1 cm cell vs water
UVat 210 nm A: =<0.7
UVat 280 nm A: =<0.05
UVat 400 nm A: =<0.01
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only