1,2-Dipiperidinoethane, 98%
1,2-Dipiperidinoethane, 98%
1,2-Dipiperidinoethane, 98%
1,2-Dipiperidinoethane, 98%
Thermo Scientific Chemicals

1,2-Dipiperidinoethane, 98%

CAS: 1932-04-3 | C12H24N2 | 196.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 185110050
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1932-04-3
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow to amber to orange
Refractive index1.4865 to 1.4885 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only