Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Thermo Scientific Chemicals

Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™

CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
檢視全部
AcroSeal™ Glass bottle
產品號碼 185451000
價格 (TWD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
化學識別
CAS563-43-9
IUPAC Namedichloro(ethyl)alumane
Molecular FormulaC2H5AlCl2
InChI KeyUAIZDWNSWGTKFZ-UHFFFAOYSA-L
SMILESCC[Al](Cl)Cl
檢視更多
規格Specification Sheet規格表
Titration Argentometric14.5 to 16.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only