Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Thermo Scientific Chemicals

Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™

CAS: 12075-68-2 | C6H15Al2Cl3 | 247.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 185468000
Price (USD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS12075-68-2
IUPAC Namechlorodiethylalumane; dichloro(ethyl)alumane
Molecular FormulaC6H15Al2Cl3
InChI KeyLDXMUMPEDOQULK-UHFFFAOYSA-K
SMILESCC[Al](Cl)Cl.CC[Al](Cl)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Assay14.5 to 15.5 % (on Al basis)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only