Borane-methyl sulfide complex, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Borane-methyl sulfide complex, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Borane-methyl sulfide complex, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Borane-methyl sulfide complex, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
185611000100 mLAcroSeal™ Glass bottle
Catalog number 185611000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13292-87-0
IUPAC Name(methylsulfanyl)methane borane
Molecular FormulaC2H9BS
InChI KeyRMHDLBZYPISZOI-UHFFFAOYSA-N
SMILESB.CSC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Strength0.95 to 1.05 M
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only