Etil nipecotato, 97 %, Thermo Scientific Chemicals
Etil nipecotato, 97 %, Thermo Scientific Chemicals
Etil nipecotato, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Etil nipecotato, 97 %, Thermo Scientific Chemicals

CAS: 5006-62-2 | C8H15NO2 | 157.21 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
500 g
Envase:
Frasco de vidrio
Número de catálogo 186621000
Precio (EUR)
94,50
Each
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
94,50
Each
Identificadores químicos
CAS5006-62-2
IUPAC Nameethyl piperidine-3-carboxylate
Molecular FormulaC8H15NO2
InChI KeyXIWBSOUNZWSFKU-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCCNC1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4580 to 1.4610 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only