1,3-Dibromobenzene, 97%
1,3-Dibromobenzene, 97%
1,3-Dibromobenzene, 97%
Thermo Scientific Chemicals

1,3-Dibromobenzene, 97%

CAS: 108-36-1 | C6H4Br2 | 235.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 186640500
Price (USD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-36-1
IUPAC Name1,3-dibromobenzene
Molecular FormulaC6H4Br2
InChI KeyJSRLURSZEMLAFO-UHFFFAOYSA-N
SMILESBrC1=CC(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.6070 to 1.6090 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only